Assay Details
| Gene Symbol: MBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116266.1 | NP_567177.1 | 1 | 339 | 93 | |
| GenBank mRNA | DQ056635.1 | - | 1 | 339 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MBD3| Gene Symbol: MBD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116266.1 | NP_567177.1 | 1 | 339 | 93 | |
| GenBank mRNA | DQ056635.1 | - | 1 | 339 | 93 | |
Set Membership: |
|