Assay Details
| Gene Symbol: BSK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340265.1 | NP_001329175.1 | 2 - 3 | 1226 | 66 | |
| NM_116296.4 | NP_191980.2 | 3 - 4 | 1238 | 66 | ||
| GenBank mRNA | AY062495.1 | - | 3 - 4 | 981 | 66 | |
| AY093275.1 | - | 2 - 3 | 301 | 66 | ||
More Information
Set Membership: |
|

BSK3
Genomic Map