Assay Details
| Gene Symbol: BSK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340265.1 | NP_001329175.1 | 3 - 4 | 1362 | 101 | |
| NM_116296.4 | NP_191980.2 | 4 - 5 | 1374 | 101 | ||
| GenBank mRNA | AY062495.1 | - | 4 - 5 | 1117 | 101 | |
| AY093275.1 | - | 3 - 4 | 437 | 101 | ||
More Information
Set Membership: |
|

BSK3
Genomic Map