Assay Details
| Gene Symbol: BSK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340265.1 | NP_001329175.1 | 7 - 8 | 1879 | 69 | |
| NM_116296.4 | NP_191980.2 | 8 - 9 | 1891 | 69 | ||
More Information
Set Membership: |
|

BSK3
Genomic Map