Assay Details
| Gene Symbol: QUA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340266.1 | NP_001328335.1 | 1 - 2 | 657 | 71 | |
| NM_116299.3 | NP_567184.1 | 1 - 2 | 624 | 71 | ||
| GenBank mRNA | AY059835.1 | - | 1 - 2 | 594 | 71 | |
| AY059913.1 | - | 1 - 2 | 594 | 71 | ||
| BT000151.1 | - | 1 - 2 | 438 | 71 | ||
| BT008377.1 | - | 1 - 2 | 438 | 71 | ||
More Information
Set Membership: |
|

QUA3
Genomic Map