Assay Details
| Gene Symbol: QUA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340266.1 | NP_001328335.1 | 6 - 7 | 1799 | 137 | |
| NM_116299.3 | NP_567184.1 | 6 - 7 | 1766 | 137 | ||
| GenBank mRNA | AY059835.1 | - | 6 - 7 | 1736 | 137 | |
| AY059913.1 | - | 6 - 7 | 1736 | 137 | ||
| BT000151.1 | - | 6 - 7 | 1580 | 137 | ||
| BT008377.1 | - | 6 - 7 | 1580 | 137 | ||
More Information
Set Membership: |
|

QUA3
Genomic Map