Assay Details
| Gene Symbol: GIF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340272.1 | NP_001328370.1 | 1 - 2 | 356 | 77 | |
| NM_116311.4 | NP_567194.1 | 1 - 2 | 408 | 77 | ||
| GenBank mRNA | AF378866.1 | - | 1 - 2 | 146 | 77 | |
| AY052732.1 | - | 1 - 2 | 78 | 77 | ||
| AY102641.1 | - | 1 - 2 | 185 | 77 | ||
More Information
Set Membership: |
|

GIF3
Genomic Map