Assay Details
Gene Symbol: GIF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001340272.1 | NP_001328370.1 | 3 - 4 | 549 | 62 | |
NM_116311.4 | NP_567194.1 | 3 - 4 | 601 | 62 | ||
GenBank mRNA | AF378866.1 | - | 3 - 4 | 339 | 62 | |
AY052732.1 | - | 3 - 4 | 271 | 62 | ||
AY102641.1 | - | 3 - 4 | 378 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
