Assay Details
| Gene Symbol: GIF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116311.4 | NP_567194.1 | 4 - 5 | 879 | 71 | |
| GenBank mRNA | AF378866.1 | - | 4 - 5 | 617 | 71 | |
| AY052732.1 | - | 4 - 5 | 549 | 71 | ||
| AY102641.1 | - | 4 - 5 | 656 | 71 | ||
More Information
Set Membership: |
|

GIF3
Genomic Map