Assay Details
| Gene Symbol: ECA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340276.1 | NP_001329712.1 | 1 - 2 | 1718 | 117 | |
| NM_116317.4 | NP_191999.1 | 2 - 3 | 1893 | 117 | ||
| GenBank mRNA | AJ132387.1 | - | 1 - 2 | 1527 | 117 | |
More Information
Set Membership: |
|

ECA2
Genomic Map