Assay Details
| Gene Symbol: AT4G00920 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116319.1 | NP_567200.1 | 5 - 6 | 734 | 86 | |
| GenBank mRNA | DQ056636.1 | - | 5 - 6 | 734 | 86 | |
More Information
Set Membership: |
|

AT4G00920
Genomic Map