Assay Details
| Gene Symbol: CRK41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340291.1 | NP_001328457.1 | 1 - 2 | 871 | 70 | |
| NM_116325.4 | NP_567204.3 | 1 - 2 | 935 | 70 | ||
More Information
Set Membership: |
|

CRK41
Genomic Map