Assay Details
| Gene Symbol: AT4G01150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125437.1 | NP_001118909.1 | 1 - 2 | 294 | 99 | |
| NM_116345.4 | NP_567210.1 | 1 - 2 | 296 | 99 | ||
| GenBank mRNA | AF389292.1 | - | 1 - 2 | 298 | 99 | |
| AY091679.1 | - | 1 - 2 | 150 | 99 | ||
More Information
Set Membership: |
|

AT4G01150
Genomic Map