Assay Details
| Gene Symbol: AT4G01150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125437.1 | NP_001118909.1 | 2 - 3 | 396 | 121 | |
| NM_116345.4 | NP_567210.1 | 2 - 3 | 398 | 121 | ||
| GenBank mRNA | AF389292.1 | - | 2 - 3 | 400 | 121 | |
| AY091679.1 | - | 2 - 3 | 252 | 121 | ||
More Information
Set Membership: |
|

AT4G01150
Genomic Map