Assay Details
| Gene Symbol: AT4G01150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001125437.1 | NP_001118909.1 | 3 - 4 | 666 | 96 | |
| NM_116345.4 | NP_567210.1 | 4 - 5 | 578 | 96 | ||
| GenBank mRNA | AF389292.1 | - | 4 - 5 | 580 | 96 | |
| AY091679.1 | - | 4 - 5 | 432 | 96 | ||
| Gene Symbol: CAM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | M38380.1 | - | 207 | 96 | ||
More Information
Set Membership: |
|

AT4G01150
Genomic Map