Assay Details
| Gene Symbol: AT4G01210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116351.7 | NP_192030.4 | 5 - 6 | 1364 | 77 | |
| GenBank mRNA | KJ138774.1 | - | 5 - 6 | 874 | 77 | |
More Information
Set Membership: |
|

AT4G01210
Genomic Map