Assay Details
| Gene Symbol: MGP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116352.3 | NP_567211.1 | 1 - 2 | 695 | 62 | |
| NM_178948.3 | NP_849279.1 | 1 - 2 | 695 | 62 | ||
| GenBank mRNA | BT020481.1 | - | 1 - 2 | 493 | 62 | |
| BT033130.1 | - | 1 - 2 | 490 | 62 | ||
| KJ138773.1 | - | 1 - 2 | 490 | 62 | ||
More Information
Set Membership: |
|

MGP4
Genomic Map