Assay Details
| Gene Symbol: ATSTE24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340307.1 | NP_001329598.1 | 10 - 11 | 1280 | 101 | |
| NM_001340308.1 | NP_001329597.1 | 11 - 12 | 1181 | 101 | ||
| NM_116362.3 | NP_567212.1 | 10 - 11 | 1015 | 101 | ||
| GenBank mRNA | AY090232.1 | - | 10 - 11 | 982 | 101 | |
| BT006355.1 | - | 10 - 11 | 906 | 101 | ||
More Information
Set Membership: |
|

ATSTE24
Genomic Map