Assay Details
| Gene Symbol: ATSTE24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340307.1 | NP_001329598.1 | 7 - 8 | 1012 | 62 | |
| NM_001340308.1 | NP_001329597.1 | 8 - 9 | 913 | 62 | ||
| NM_116362.3 | NP_567212.1 | 7 - 8 | 747 | 62 | ||
| GenBank mRNA | AY090232.1 | - | 7 - 8 | 714 | 62 | |
| BT006355.1 | - | 7 - 8 | 638 | 62 | ||
More Information
Set Membership: |
|

ATSTE24
Genomic Map