Assay Details
| Gene Symbol: ATSTE24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340307.1 | NP_001329598.1 | 8 - 9 | 1125 | 110 | |
| NM_001340308.1 | NP_001329597.1 | 9 - 10 | 1026 | 110 | ||
| NM_116362.3 | NP_567212.1 | 8 - 9 | 860 | 110 | ||
| GenBank mRNA | AY090232.1 | - | 8 - 9 | 827 | 110 | |
| BT006355.1 | - | 8 - 9 | 751 | 110 | ||
More Information
Set Membership: |
|

ATSTE24
Genomic Map