Assay Details
| Gene Symbol: MPK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116367.3 | NP_192046.1 | 1 - 2 | 476 | 63 | |
| GenBank mRNA | AF360231.1 | - | 1 - 2 | 223 | 63 | |
| AY040031.1 | - | 1 - 2 | 174 | 63 | ||
| AY088537.1 | - | 1 - 2 | 298 | 63 | ||
More Information
Set Membership: |
|

MPK4
Genomic Map