Assay Details
| Gene Symbol: CRK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116401.3 | NP_567216.1 | 2 - 3 | 289 | 66 | |
| GenBank mRNA | AY052191.1 | - | 2 - 3 | 189 | 66 | |
| AY143806.1 | - | 2 - 3 | 166 | 66 | ||
More Information
Set Membership: |
|

CRK
Genomic Map