Assay Details
| Gene Symbol: KCO5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116414.4 | NP_192093.1 | 1 - 2 | 1300 | 110 | |
| GenBank mRNA | AJ243456.1 | - | 1 - 2 | 1075 | 110 | |
More Information
Set Membership: |
|

KCO5
Genomic Map