Assay Details
| Gene Symbol: PRL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203726.1 | NP_001190655.1 | 6 - 7 | 545 | 117 | |
| NM_116437.3 | NP_192115.1 | 4 - 5 | 495 | 117 | ||
More Information
Set Membership: |
|

PRL
Genomic Map