Assay Details
| Gene Symbol: SYP42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116452.4 | NP_567223.1 | 1 - 2 | 339 | 91 | |
| GenBank mRNA | AF154574.1 | - | 1 - 2 | 189 | 91 | |
| AK229319.1 | - | 1 - 2 | 301 | 91 | ||
More Information
Set Membership: |
|

SYP42
Genomic Map