Assay Details
| Gene Symbol: SYP42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116452.4 | NP_567223.1 | 3 - 4 | 708 | 105 | |
| GenBank mRNA | AF154574.1 | - | 3 - 4 | 558 | 105 | |
| AK229319.1 | - | 3 - 4 | 670 | 105 | ||
More Information
Set Membership: |
|

SYP42
Genomic Map