Assay Details
| Gene Symbol: SR34b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340375.1 | NP_001319845.1 | 4 - 5 | 593 | 134 | |
| NM_001340376.1 | NP_001328195.1 | 5 - 6 | 408 | 134 | ||
| NM_001340377.1 | NP_001328194.1 | 5 - 6 | 408 | 134 | ||
| NM_179206.3 | NP_849537.1 | 5 - 6 | 479 | 134 | ||
| GenBank mRNA | AK118074.1 | - | 5 - 6 | 512 | 134 | |
More Information
Set Membership: |
|

SR34b
Genomic Map