Assay Details
| Gene Symbol: MLO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036501.2 | NP_001031578.1 | 4 - 5 | 727 | 74 | |
| NM_116494.5 | NP_192169.1 | 3 - 4 | 811 | 74 | ||
| GenBank mRNA | AY072135.1 | - | 3 - 4 | 658 | 74 | |
| AY113992.1 | - | 3 - 4 | 558 | 74 | ||
| Z95352.1 | - | 650 | 74 | |||
More Information
Set Membership: |
|

MLO1
Genomic Map