Assay Details
| Gene Symbol: MLO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001036501.2 | NP_001031578.1 | 7 - 8 | 960 | 131 | |
| NM_116494.5 | NP_192169.1 | 6 - 7 | 1044 | 131 | ||
| GenBank mRNA | AY072135.1 | - | 6 - 7 | 891 | 131 | |
| AY113992.1 | - | 6 - 7 | 791 | 131 | ||
| Z95352.1 | - | 883 | 131 | |||
More Information
Set Membership: |
|

MLO1
Genomic Map