Assay Details
| Gene Symbol: SULTR3;2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116504.3 | NP_192179.1 | 5 - 6 | 960 | 122 | |
| GenBank mRNA | AB004060.1 | - | 5 - 6 | 920 | 122 | |
| AK229084.1 | - | 5 - 6 | 960 | 122 | ||
More Information
Set Membership: |
|

SULTR3;2
Genomic Map