Assay Details
| Gene Symbol: WDR5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116507.3 | NP_192182.1 | 1 - 2 | 789 | 128 | |
| GenBank mRNA | AF439825.1 | - | 1 - 2 | 745 | 128 | |
| AY088121.1 | - | 1 - 2 | 748 | 128 | ||
| AY125558.1 | - | 1 - 2 | 713 | 128 | ||
More Information
Set Membership: |
|

WDR5b
Genomic Map