Assay Details
| Gene Symbol: GRH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116555.4 | NP_567255.1 | 1 - 2 | 798 | 64 | |
| GenBank mRNA | AY150427.1 | - | 1 - 2 | 453 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRH1| Gene Symbol: GRH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116555.4 | NP_567255.1 | 1 - 2 | 798 | 64 | |
| GenBank mRNA | AY150427.1 | - | 1 - 2 | 453 | 64 | |
Set Membership: |
|