Assay Details
| Gene Symbol: SYP123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116571.3 | NP_192242.1 | 1 - 2 | 724 | 89 | |
| GenBank mRNA | AK229042.1 | - | 1 - 2 | 566 | 89 | |
| AY087693.1 | - | 1 - 2 | 672 | 89 | ||
| BT044612.1 | - | 1 - 2 | 483 | 89 | ||
More Information
Set Membership: |
|

SYP123
Genomic Map