Assay Details
| Gene Symbol: SRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340445.1 | NP_001329389.1 | 9 - 10 | 1382 | 68 | |
| NM_116577.5 | NP_192248.2 | 10 - 11 | 1304 | 68 | ||
| GenBank mRNA | AY518288.1 | - | 10 - 11 | 977 | 68 | |
| FJ708743.1 | - | 9 - 10 | 721 | 68 | ||
More Information
Set Membership: |
|

SRF3
Genomic Map