Assay Details
| Gene Symbol: SRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340445.1 | NP_001329389.1 | 10 - 11 | 1815 | 63 | |
| NM_116577.5 | NP_192248.2 | 11 - 12 | 1737 | 63 | ||
| GenBank mRNA | AY518288.1 | - | 11 - 12 | 1410 | 63 | |
| FJ708743.1 | - | 10 - 11 | 1154 | 63 | ||
More Information
Set Membership: |
|

SRF3
Genomic Map