Assay Details
| Gene Symbol: SRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340445.1 | NP_001329389.1 | 15 - 16 | 2883 | 129 | |
| NM_116577.5 | NP_192248.2 | 16 - 17 | 2805 | 129 | ||
| GenBank mRNA | AY518288.1 | - | 16 - 17 | 2478 | 129 | |
| FJ708743.1 | - | 15 - 16 | 2222 | 129 | ||
More Information
Set Membership: |
|

SRF3
Genomic Map