Assay Details
| Gene Symbol: SRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340445.1 | NP_001329389.1 | 1 - 2 | 765 | 70 | |
| NM_116577.5 | NP_192248.2 | 2 - 3 | 687 | 70 | ||
| GenBank mRNA | AY518288.1 | - | 2 - 3 | 360 | 70 | |
| FJ708743.1 | - | 1 - 2 | 104 | 70 | ||
More Information
Set Membership: |
|

SRF3
Genomic Map