Assay Details
| Gene Symbol: SRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340445.1 | NP_001329389.1 | 3 - 4 | 973 | 97 | |
| NM_116577.5 | NP_192248.2 | 4 - 5 | 895 | 97 | ||
| GenBank mRNA | AY518288.1 | - | 4 - 5 | 568 | 97 | |
| FJ708743.1 | - | 3 - 4 | 312 | 97 | ||
More Information
Set Membership: |
|

SRF3
Genomic Map