Assay Details
| Gene Symbol: SRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340445.1 | NP_001329389.1 | 5 - 6 | 1110 | 78 | |
| NM_116577.5 | NP_192248.2 | 6 - 7 | 1032 | 78 | ||
| GenBank mRNA | AY518288.1 | - | 6 - 7 | 705 | 78 | |
| FJ708743.1 | - | 5 - 6 | 449 | 78 | ||
More Information
Set Membership: |
|

SRF3
Genomic Map