Assay Details
| Gene Symbol: SRF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340445.1 | NP_001329389.1 | 7 - 8 | 1257 | 95 | |
| NM_116577.5 | NP_192248.2 | 8 - 9 | 1179 | 95 | ||
| GenBank mRNA | AY518288.1 | - | 8 - 9 | 852 | 95 | |
| FJ708743.1 | - | 7 - 8 | 596 | 95 | ||
More Information
Set Membership: |
|

SRF3
Genomic Map