Assay Details
| Gene Symbol: PFA-DSP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116634.4 | NP_567261.1 | 3 - 4 | 557 | 85 | |
| GenBank mRNA | AF458342.1 | - | 3 - 4 | 346 | 85 | |
| AK118704.1 | - | 3 - 4 | 377 | 85 | ||
| AY054279.1 | - | 3 - 4 | 490 | 85 | ||
| AY086713.1 | - | 3 - 4 | 411 | 85 | ||
More Information
Set Membership: |
|

PFA-DSP4
Genomic Map