Assay Details
| Gene Symbol: AT4G04340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116672.4 | NP_192343.1 | 10 - 11 | 2254 | 107 | |
| NM_178965.2 | NP_849296.1 | 10 - 11 | 2108 | 107 | ||
| NM_178966.1 | NP_849297.1 | 9 - 10 | 2371 | 107 | ||
| GenBank mRNA | AY093209.1 | - | 10 - 11 | 2073 | 107 | |
| KJ920356.1 | - | 9 - 10 | 1824 | 107 | ||
More Information
Set Membership: |
|

AT4G04340
Genomic Map