Assay Details
| Gene Symbol: AT4G04340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116672.4 | NP_192343.1 | 3 - 4 | 904 | 133 | |
| NM_178965.2 | NP_849296.1 | 3 - 4 | 758 | 133 | ||
| NM_178966.1 | NP_849297.1 | 2 - 3 | 1021 | 133 | ||
| GenBank mRNA | AY093209.1 | - | 3 - 4 | 723 | 133 | |
| KJ920356.1 | - | 2 - 3 | 474 | 133 | ||
More Information
Set Membership: |
|

AT4G04340
Genomic Map