Assay Details
| Gene Symbol: AT4G04340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116672.4 | NP_192343.1 | 6 - 7 | 1265 | 111 | |
| NM_178965.2 | NP_849296.1 | 6 - 7 | 1119 | 111 | ||
| NM_178966.1 | NP_849297.1 | 5 - 6 | 1382 | 111 | ||
| GenBank mRNA | AY093209.1 | - | 6 - 7 | 1084 | 111 | |
| KJ920356.1 | - | 5 - 6 | 835 | 111 | ||
More Information
Set Membership: |
|

AT4G04340
Genomic Map