Assay Details
| Gene Symbol: AT4G04340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116672.4 | NP_192343.1 | 8 - 9 | 1685 | 71 | |
| NM_178965.2 | NP_849296.1 | 8 - 9 | 1539 | 71 | ||
| NM_178966.1 | NP_849297.1 | 7 - 8 | 1802 | 71 | ||
| GenBank mRNA | AY093209.1 | - | 8 - 9 | 1504 | 71 | |
| KJ920356.1 | - | 7 - 8 | 1255 | 71 | ||
More Information
Set Membership: |
|

AT4G04340
Genomic Map