Assay Details
| Gene Symbol: CRK40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084882.2 | NP_001078351.1 | 4 - 5 | 1449 | 121 | |
| NM_001340491.1 | NP_001329203.1 | 5 - 6 | 1716 | 121 | ||
| NM_116695.3 | NP_192366.1 | 4 - 5 | 1449 | 121 | ||
| GenBank mRNA | AY127017.1 | - | 4 - 5 | 1392 | 121 | |
More Information
Set Membership: |
|

CRK40
Genomic Map