Assay Details
| Gene Symbol: AT4G04670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116705.5 | NP_567268.2 | 5 - 6 | 2737 | 88 | |
| GenBank mRNA | AY062510.1 | - | 5 - 6 | 2605 | 88 | |
| BT008367.1 | - | 5 - 6 | 2568 | 88 | ||
More Information
Set Membership: |
|

AT4G04670
Genomic Map