Assay Details
| Gene Symbol: AT4G04670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116705.5 | NP_567268.2 | 7 - 8 | 2958 | 66 | |
| GenBank mRNA | AY062510.1 | - | 7 - 8 | 2826 | 66 | |
| BT008367.1 | - | 7 - 8 | 2789 | 66 | ||
More Information
Set Membership: |
|

AT4G04670
Genomic Map