Assay Details
| Gene Symbol: MED21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116716.5 | NP_192387.2 | 3 - 4 | 473 | 70 | |
| GenBank mRNA | FJ769239.1 | - | 2 - 3 | 342 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MED21| Gene Symbol: MED21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_116716.5 | NP_192387.2 | 3 - 4 | 473 | 70 | |
| GenBank mRNA | FJ769239.1 | - | 2 - 3 | 342 | 70 | |
Set Membership: |
|