Assay Details
| Gene Symbol: RIC10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340518.1 | NP_001319870.1 | 3 - 4 | 637 | 61 | |
| NM_116728.2 | NP_192399.1 | 2 - 3 | 191 | 61 | ||
More Information
Set Membership: |
|

RIC10
Genomic Map